missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MS4A12 Antibody (OTI5F5), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-72791CL1
This item is not returnable.
View return policy
Description
MS4A12 Monoclonal antibody specifically detects MS4A12 in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| MS4A12 | |
| Monoclonal | |
| CoraFluor 1 | |
| FLJ20217, membrane-spanning 4-domains subfamily A member 12, membrane-spanning 4-domains, subfamily A, member 12, Ms4a10 | |
| Full length human recombinant protein of human MS4A12 (NP_060186) produced in HEK293T cell. | |
| 0.1 mL | |
| Cell Biology | |
| 54860 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Western Blot | |
| OTI5F5 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction