missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MS4A12 Antibody (OTI5F5), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72791AF647
This item is not returnable.
View return policy
Description
MS4A12 Monoclonal specifically detects MS4A12 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| MS4A12 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MS4A12 | |
| Full length recombinant protein of human MS4A12 (NP_060186) produced in HEK293T cells. | |
| 0.1 mL | |
| 54860.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI5F5 | |
| Western Blot, Immunohistochemistry | |
| FLJ20217, membrane-spanning 4-domains subfamily A member 12, membrane-spanning 4-domains, subfamily A, member 12, Ms4a10 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction