missing translation for 'onlineSavingsMsg'
Learn More

MS4A10 Antibody (PSH0-71), Novus Biologicals™

Product Code. 30113501 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30113501 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30113501 Supplier Novus Biologicals Supplier No. NBP332603

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

MS4A10 Monoclonal antibody specifically detects MS4A10 in Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen MS4A10
Applications Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PSH0-71
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:100-1:500
Formulation PBS (pH7.4), 0.1% BSA and 40% Glycerol
Gene Alias CD20L7, membrane-spanning 4-domains, subfamily A, member 10, MS4A9
Host Species Rabbit
Immunogen Synthetic peptide within mouse MS4A10 aa 218-267/ 267. (Uniprot: Q99N03)
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 341116
Target Species Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.