missing translation for 'onlineSavingsMsg'
Learn More

MS436, MedChemExpress

Product Code. 16428567
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16428567

Brand: Medchem Express HY13959

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MS436 is a new class of bromodomain inhibitor, exhibits potent affinity of an estimated Ki=30-50 nM for the BRD4 BrD1 and a 10-fold selectivity over the BrD2.

MS436 is a new class of bromodomain inhibitor, exhibits potent affinity of an estimated Ki=30-50 nM for the BRD4 BrD1 and a 10-fold selectivity over the BrD2.

Specifications

Chemical Name or Material MS436
CAS 1395084-25-9
Color Reddish Brown
Purity Grade Notes Research
Molecular Formula C18H17N5O3S
Quantity 5 mg
Solubility Information DMSO : 25.5 mg/mL (66.51 mM; Need ultrasonic and warming)
SMILES O=S(C1=CC=C(/N=N/C2=CC(C)=C(O)C=C2N)C=C1)(NC3=NC=CC=C3)=O
Molecular Weight (g/mol) 383.42
Formula Weight 383.42
Percent Purity 98.07%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.