missing translation for 'onlineSavingsMsg'
Learn More

MS417, MedChemExpress

Product Code. p-200099664
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16494408

Brand: Medchem Express HY111139

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MS417 is a selective BET-specific BRD4 inhibitor, binds to BRD4-BD1 and BRD4-BD2 with IC50s of 30, 46 nM and Kds of 36.1, 25.4 nM, respectively, with weak selectivity at CBP BRD (IC50, 32.7 μM).

MS417 is a selective BET-specific BRD4 inhibitor, binds to BRD4-BD1 and BRD4-BD2 with IC50s of 30, 46 nM and Kds of 36.1, 25.4 nM, respectively, with weak selectivity at CBP BRD (IC50, 32.7 μM).

Specifications

Chemical Name or Material MS417
CAS 916489-36-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C20H19ClN4O2S
Quantity 10 mg
Synonym GTPL7512
Solubility Information Ethanol : 50 mg/mL (120.51 mM; Need ultrasonic)
SMILES O=C(OC)C[C@H]1C2=NN=C(C)N2C3=C(C(C)=C(C)S3)C(C4=CC=C(Cl)C=C4)=N1
Molecular Weight (g/mol) 414.91
Formula Weight 414.91
Percent Purity 96.47%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.