missing translation for 'onlineSavingsMsg'
Learn More

MS417, MedChemExpress

Product Code. 16494408
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494408 10 mg 10mg
16470727 50 mg 50mg
16445306 100 mg 100mg
16407137 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16494408 Supplier Medchem Express Supplier No. HY111139

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MS417 is a selective BET-specific BRD4 inhibitor, binds to BRD4-BD1 and BRD4-BD2 with IC50s of 30, 46 nM and Kds of 36.1, 25.4 nM, respectively, with weak selectivity at CBP BRD (IC50, 32.7 μM).

MS417 is a selective BET-specific BRD4 inhibitor, binds to BRD4-BD1 and BRD4-BD2 with IC50s of 30, 46 nM and Kds of 36.1, 25.4 nM, respectively, with weak selectivity at CBP BRD (IC50, 32.7 μM).

Specifications

Chemical Name or Material MS417
CAS 916489-36-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C20H19ClN4O2S
Quantity 10 mg
Synonym GTPL7512
Solubility Information Ethanol : 50 mg/mL (120.51 mM; Need ultrasonic)
SMILES O=C(OC)C[C@H]1C2=NN=C(C)N2C3=C(C(C)=C(C)S3)C(C4=CC=C(Cl)C=C4)=N1
Molecular Weight (g/mol) 414.91
Formula Weight 414.91
Percent Purity 96.47%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.