missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MS049, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Description
MS049 is a potent, selective, and cell-active dual inhibitor of PRMT4 and PRMT6 with IC50s of 34 nM and 43 nM, respectively. MS049 reduces levels of Med12me2a and H3R2me2a in HEK293 cells. MS049 is not toxic and does not affect the growth of HEK293 cells.
Specifications
Specifications
| Chemical Name or Material | MS049 |
| Physical Form | Solid |
| CAS | 1502816-23-0 |
| Color | Light Yellow |
| Quantity | 25 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C15H24N2O |
| For Use With (Application) | Cancer-programmed cell death |
| Solubility Information | H2O : ≥ 100 mg/mL (402.64 mM) ∣DMSO : ≥ 31 mg/mL (124.82 mM) |
| SMILES | CNCCN1CCC(OCC2=CC=CC=C2)CC1 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction