missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ MS 436
Description
MS 436 is a potent and selective BRD4 bromodomain inhibitor (Ki = 30 - 50 nM for the first bromodomain (BRD4(1)). Exhibits 10-fold selectivity for BRD4(1) over BRD4(2). Blocks BRD4 transcriptional activity in lipopolysaccharide-induced production of both nitric oxide and IL-6 in mouse macrophages (IC50 values are 3.8 and 4.9 μM, respectively). Attenuates melanoma cell proliferation in vitro.
Specifications
Specifications
| Purity | >98% |
| Quantity | 10 mg |
| Product Type | MS 436 |
| Molecular Weight (g/mol) | 383.42 |
| Inhibitors | MS 436 |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?