missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ MS 154

Product Code. 18797856
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18797856 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18797856 Supplier R&D Systems™ Supplier No. 7395/5
  • ONLINE EXCLUSIVE PRICE
    £328.00
    £309.75 / 5mg
    Save £18.25
    5% Off
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective cereblon-recruiting Degrader (PROTAC™) of mutant EGFR

MS 154 is a potent and selective cereblon-recruiting Degrader (PROTAC™) of mutant epidermal growth factor receptor (EGFR), comprising a cereblon-binding moiety joined by a linker to gefitinib (Iressa, Cat. No. 3000). MS 154 decreases EGFR protein levels, inhibits downstream signaling in and inhibits proliferation of mutant EGFR-bearing lung cancer cells (DC50 values are 11 and 25 nM in HCC-827 and H3255 cells, respectively; Dmax > 95% at 50 nM), but not in WT-EGFR-bearing ovarian and lung cancer cells lines. Bioavailable in mice following ip administration.

Negative control (Cat. No. 7396) also available.

PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(4-(3-((4-((3-Chloro-4-fluorophenyl)amino)-7-methoxyquinazolin-6-yl)oxy)propyl)piperazin-1-yl)-N-(8-((2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)oxy)octyl)propanamide
CAS 2550393-21-8
Quantity 5 mg
Product Line Tocris
Target Active Degraders
Molecular Formula C46H54ClFN8O8
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.