missing translation for 'onlineSavingsMsg'
Learn More

MRX-2843, MedChemExpress

Product Code. p-200100418
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16492756

Brand: Medchem Express HY101549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MRX-2843 (UNC2371) is an orally active, ATP-competitive dual MERTK and FLT3 tyrosine kinases inhibitor (TKI) with enzymatic IC50s of 1.3 nM for MERTK and 0.64 nM for FLT3, respectively.

MRX-2843 (UNC2371) is an orally active, ATP-competitive dual MERTK and FLT3 tyrosine kinases inhibitor (TKI) with enzymatic IC50s of 1.3 nM for MERTK and 0.64 nM for FLT3, respectively.

Specifications

Chemical Name or Material MRX-2843
CAS 1429882-07-4
Color Light Brown
Purity Grade Notes Research
Molecular Formula C29H40N6O
Quantity 25 mg
Synonym UNC2371
Solubility Information DMSO : 31.25 mg/mL (63.95 mM; Need ultrasonic)
SMILES O[C@H]1CC[C@H](N2C=C(C3=CC=C(CN4CCN(C)CC4)C=C3)C5=CN=C(NCCC6CC6)N=C52)CC1
Molecular Weight (g/mol) 488.67
Formula Weight 488.67
Percent Purity 98.09%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.