missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRTO4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00347-100ul
This item is not returnable.
View return policy
Description
MRTO4 Polyclonal antibody specifically detects MRTO4 in Human samples. It is validated for ELISA
Specifications
| MRTO4 | |
| Polyclonal | |
| ELISA 1:1000-1:2000 | |
| C1orf33, chromosome 1 open reading frame 33, dJ657E11.4, mRNA turnover 4 homolog (S. cerevisiae), mRNA turnover protein 4 homolog, MRT4, mRNA turnover 4, homolog, MRT4, mRNA turnover 4, homolog (S. cerevisiae), MRT460S acidic ribosomal protein PO | |
| Produced in rabbits immunized with purified, recombinant Human MRTO4 (Uniprot#: Q9UKD2; Met1-Asp239) | |
| 100 μg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 51154 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction