missing translation for 'onlineSavingsMsg'
Learn More

MRT67307, MedChemExpress

Product Code. 16498197
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498197 5 mg 5mg
16445488 10 mg 10mg
16416076 100 mg 100mg
16401667 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16498197 Supplier Medchem Express Supplier No. HY13018

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MRT67307 is a dual inhibitor of the IKKε and TBK-1 with IC50s of 160 and 19 nM, respectively. MRT67307 also inhibits ULK1 and ULK2 with IC50s of 45 and 38 nM, respectively. MRT67307 also blocks autophagy in cells.

MRT67307 is a dual inhibitor of the IKKε and TBK-1 with IC50s of 160 and 19 nM, respectively. MRT67307 also inhibits ULK1 and ULK2 with IC50s of 45 and 38 nM, respectively. MRT67307 also blocks autophagy in cells.

Specifications

Chemical Name or Material MRT67307
Physical Form Solid
CAS 1190378-57-4
Color Light Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C26H36N6O2
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 100 mg/mL (215.24 mM)
SMILES O=C(NCCCNC1=NC(NC2=CC=CC(CN3CCOCC3)=C2)=NC=C1C4CC4)C5CCC5
Molecular Weight (g/mol) 464.6
Formula Weight 464.6
Percent Purity 98.76%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.