missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRS2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553616
This item is not returnable.
View return policy
Description
Immunogen sequence: NTLQGKLSIL QPLILETLDA LVDPKHSSVD RSKLHILLQN GKSLSELETD IKIFKESILE ILDEEELLEE LCVSKWSDPQ VFEKSSAGID HAEEMELLLE NYYRLADDLS NAARELRVLI DDSQSIIFI Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 93%.
Mrs2 is a magnesium transporter that mediates the influx of magnesium into the mitochondrial matrix. Mrs2 is required for normal expression of the mitochondrial respiratory complex I subunits, and mediates the influx of magnesium into the mitochondrial matrix. Mrs2 is also required for normal expression of the mitochondrial respiratory complex I subunits.
Specifications
| MRS2 | |
| Polyclonal | |
| Unconjugated | |
| MRS2 | |
| Gm902; HPT; Magnesium transporter MRS2 homolog, mitochondrial; MRS2; MRS2 magnesium homeostasis factor homolog; MRS2 magnesium homeostasis factor homolog (S. cerevisiae); MRS2 magnesium transporter; MRS2, magnesium transporter; MRS2L; MRS2-like protein; MRS2-like, magnesium homeostasis factor; putative magnesium transporter; RPT; RPT protein similar to yeast MRS2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57380 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9HD23 | |
| MRS2 | |
| Recombinant protein corresponding to Human MRS2. Recombinant protein control fragment (Product #RP-90946). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction