missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ MRS 2693 trisodium salt

Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Selective P2Y6 agonist (EC50 = 0.015 μM at the hP2Y6 receptor). Displays no activity at other P2Y subtypes. Exerts a cytoprotective effect in an in vivo mouse model of hindlimb skeletal muscle ischemia/reperfusion injury.
Specifications
Specifications
| CAS | 1448858-83-0 |
| Quantity | 1 mg |
| Molecular Formula | C9H13IN2Na3O12P2 |
| Formula Weight | 596 |
| Purity | >95% |
| Molecular Weight (g/mol) | 599.026 |
| Chemical Name or Material | MRS 2693 trisodium salt |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction