Learn More
Tocris Bioscience™ MRS 2578
GREENER_CHOICE
Product Code. 18745586
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 18745586
Brand: Tocris Bioscience™ 2146/10
Selective P2Y6 antagonist
MRS 2578 is a selective antagonist of P2Y6 nucleotide receptors; IC50 values are 37 and 98 nM at human and rat P2Y6 receptors respectively. Displays no activity at P2Y1, P2Y2, P2Y4 and P2Y11 receptors (IC50 > 10 μM). Inhibits agonist-induced cardiomyocyte contraction and UDP-induced phagocytosis.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10 mg
Product Type MRS 2578
Molecular Weight (g/mol) 472.66
Inhibitors MRS 2578