missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ MRS 2279
GREENER_CHOICE

Product Code. 18094822
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 18094822

Brand: Tocris Bioscience™ 2158/1

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Selective, high affinity P2Y1 antagonist

MRS 2279 is a selective, high affinity competitive antagonist of the P2Y1 receptor (Ki = 2.5 nM; IC50 = 51.6 nM). Fails to block nucleotide signaling at most other P2Y receptors (P2Y2, P2Y4, P2Y6, P2Y11 and P2Y12) and potently inhibits ADP-induced aggregation of human blood platelets in vitro (pKB = 8.05).
TRUSTED_SUSTAINABILITY

Specifications

Purity >98%
CAS 2387505-47-5
Quantity 1 mg
Product Type MRS 2279
Molecular Weight (g/mol) 503.78
Inhibitors MRS 2279

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.