missing translation for 'onlineSavingsMsg'
Learn More

MRRF Antibody (7D10), DyLight 405, Novus Biologicals™

Product Code. 18421618 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18421618 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18421618 Supplier Novus Biologicals Supplier No. NBP222564V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MRRF Monoclonal antibody specifically detects MRRF in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRRF
Applications Western Blot, ELISA
Classification Monoclonal
Clone 7D10
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias mitochondrial, mitochondrial ribosome recycling factor, MTRRF
Host Species Mouse
Immunogen Recombinant human MRRF (56-262aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 92399
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.