missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRRF Antibody (7D10), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-22564CL1
This item is not returnable.
View return policy
Description
MRRF Monoclonal antibody specifically detects MRRF in Human samples. It is validated for ELISA,Western Blot
Specifications
| MRRF | |
| Monoclonal | |
| CoraFluor 1 | |
| mitochondrial, mitochondrial ribosome recycling factor, MTRRF | |
| Recombinant human MRRF (56-262aa) purified from E. coli | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 92399 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| ELISA, Western Blot | |
| 7D10 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction