missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538954
This item is not returnable.
View return policy
Description
Mammalian mitochondrial ribosomes (mitoribosomes) are responsible for protein synthesis within the mitochondrion. The mitoribosomes are composed of a 4:1 ratio of protein to RNA, with the proteins forming two subunits, the 28S subunit and the 39S subunit. Across species, the proteins that make up the mitoribosome subunits vary greatly in sequence, preventing easy recognition by sequence homology. MRP-S9 (mitochondrial 28S ribosomal protein S9), also known as RPMS9, is a 396 amino acid mitochondrial ribosomal protein. Localized to mitochondria, MRP-S9 is present in the 28S subunit of the mitoribosomes, which comprises a 12S rRNA and at least 30 distinct proteins.
Specifications
| MRPS9 | |
| Polyclonal | |
| Unconjugated | |
| MRPS9 | |
| 28S ribosomal protein S9, mitochondrial, MRP-S9, S9mt, MRPS9, RPMS9 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 301371, 64965, 69527 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P82933, Q9D7N3 | |
| MRPS9 | |
| A synthetic peptide derived from the internal region of human MRPS9 | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction