missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS9 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£149.00 - £366.00
Specifications
| Antigen | MRPS9 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:100-1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18655581
|
Novus Biologicals
NBP2-93709-0.02ml |
0.02 mL |
£149.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18655011
|
Novus Biologicals
NBP2-93709-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MRPS9 Polyclonal antibody specifically detects MRPS9 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| MRPS9 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Endocrinology | |
| PBS (pH 7.3), 50% glycerol | |
| 64965 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:100-1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| mitochondrial ribosomal protein S9, MRP-S9RPMS9S9mt28S ribosomal protein S9, mitochondrial | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 237-396 of human MRPS9 (NP_872578.1). EEEFVQRFRRSVTLESKKQLIEPVQYDEQGMAFSKSEGKRKTAKAEAIVYKHGSGRIKVNGIDYQLYFPITQDREQLMFPFHFVDRLGKHDVTCTVSGGGRSAQAGAIRLAMAKALCSFVTEDEVEWMRQAGLLTTDPRVRERKKPGQEGARRKFTWKKR | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title