missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPS7 Monoclonal Antibody (OTI5E11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526588
This item is not returnable.
View return policy
Description
MRPS7 Monoclonal Antibody for Western Blot
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein. In the prokaryotic ribosome, the comparable protein is thought to play an essential role in organizing the 3' domain of the 16 S rRNA in the vicinity of the P- and A-sites. Pseudogenes corresponding to this gene are found on chromosomes 8p and 12p.
Specifications
| MRPS7 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9Y2R9 | |
| MRPS7 | |
| Human recombinant protein fragment corresponding to amino acids 38-242 of MRPS7 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI5E11 | |
| Unconjugated | |
| MRPS7 | |
| 28S ribosomal protein S7, mitochondrial; 30S ribosomal protein S7 homolog; bMRP27a; bMRP-27a; mitchondrial ribosomal protein S7; mitochondrial ribosomal protein S7; Mitochondrial small ribosomal subunit protein uS7m; MRP-S; Mrps7; MRP-S7; ribosomal protein, mitochondrial, S7; Rpms7; RP-S7; S7mt | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 51081 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction