missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS6 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00064968-B01P
This item is not returnable.
View return policy
Description
MRPS6 Polyclonal antibody specifically detects MRPS6 in Human samples. It is validated for Western Blot
Specifications
| MRPS6 | |
| Polyclonal | |
| Western Blot 1:500 | |
| NP_115865.1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.4) | |
| C21orf101, mitochondrial ribosomal protein S6, MRP-S6chromosome 21 open reading frame 101, RPMS628S ribosomal protein S6, mitochondrial, S6mt | |
| MRPS6 (NP_115865.1, 1 a.a. - 125 a.a.) full-length human protein. MPRYELALILKAMQRPETAATLKRTIEALMDRGAIVRDLENLGERALPYRISAHSQQHNRGGYFLVDFYAPTAAVESMVEHLSRDIDVIRGNIVKHPLTQELKECEGIVPVPLAEKLYSTKKRKK | |
| 0.05 mg | |
| Endocrinology | |
| 64968 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction