missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPS36 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101671
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MRPS36.
ROD1 is a functional homolog of nrd1, an S. pombe RNA-binding protein that suppresses the onset of differentiation.
Specifications
| MRPS36 | |
| Polyclonal | |
| Unconjugated | |
| MRPS36 | |
| 1110018B13Rik; 28S ribosomal protein S36, mitochondrial; DC47; mitochondrial ribosomal protein S36; Mrps36; MRP-S36; RT36; S36mt | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 66128, 92259 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P82909, Q9CQX8 | |
| MRPS36 | |
| A synthesized peptide derived from human MRPS36(Accession P82909), corresponding to amino acid residues M6-S56. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction