missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS36 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98135-100ul
This item is not returnable.
View return policy
Description
MRPS36 Polyclonal antibody specifically detects MRPS36 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| MRPS36 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| DC47,28S ribosomal protein S36, mitochondrial, MGC22896, mitochondrial ribosomal protein S36, MRP-S36S36mt | |
| Produced in rabbits immunized with E. coli-derived Human MRPS36 fragment. | |
| 100 μg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 92259 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction