missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS35 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98524-50ul
This item is not returnable.
View return policy
Description
MRPS35 Polyclonal antibody specifically detects MRPS35 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| MRPS35 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-5 μL/mg of lysate, Immunohistochemistry-Paraffin 1:50-1:200 | |
| 28S ribosomal protein S28, mitochondrial, DKFZp762P093, MDS023,28S ribosomal protein S35, mitochondrial, MGC104278, mitochondrial ribosomal protein S28, mitochondrial ribosomal protein S35, MRP-S28, MRPS28S35mt, MRP-S35, S28mt | |
| Produced in rabbits immunized with E. coli-derived Human MRPS35 fragment. | |
| 50 μg | |
| Endocrinology | |
| 60488 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction