missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS35 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94003-0.02ml
This item is not returnable.
View return policy
Description
MRPS35 Polyclonal antibody specifically detects MRPS35 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| MRPS35 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin | |
| 28S ribosomal protein S28, mitochondrial, DKFZp762P093, MDS023,28S ribosomal protein S35, mitochondrial, MGC104278, mitochondrial ribosomal protein S28, mitochondrial ribosomal protein S35, MRP-S28, MRPS28S35mt, MRP-S35, S28mt | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-323 of human MRPS35 (NP_068593.2). MAAAALPAWLSLQSRARTLRAFSTAVYSATPVPTPSLPERTPGNERPPRRKALPPRTEKMAVDQDWPSVYPVAAPFKPSAVPLPVRMGYPVKKGVPMAKEGNLELLKIPNFLHLTPVAIKKHCEALKDFCTEWPAALDSDEKCEKHFPIEIDSTDYVSSGPSVRNPRARVVVLRVKLSSLNLDDHAKKKLIKLVGERYCKTTDVLTIKTDRCPLRRQNYDYAVYLLTVLYHESWNTEEWEKSKTEADMEEYIWENSSSERNILETLLQMKAAEKNMEINKEELLGTKEIEEYKKSVVSLKNEEENENSISQYKESVKRLLNVT | |
| 0.02 mL | |
| Endocrinology | |
| 60488 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction