missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPS34 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559872
This item is not returnable.
View return policy
Description
Immunogen sequence: PEDSLASVPY PPLLRAMIIA ERQKNGDTST EEPMLNVQRI RMEPWDYPAK QEDKGRAKGT Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 80%.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein. Alternate splice variants of this gene have been described but their full-length nature has not been determined.
Specifications
| MRPS34 | |
| Polyclonal | |
| Unconjugated | |
| MRPS34 | |
| 0610007F04Rik; 28S ribosomal protein S34, mitochondrial; 5330430D13Rik; AV001970; mitochondrial 28S ribosomal protein S34; mitochondrial ribosomal protein S34; Mitochondrial small ribosomal subunit protein mS34; MRPS12; MRP-S12; MRPS34; MRP-S34; rt34; S34mt; Tce2; T-complex expressed gene 2 protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 287126, 65993, 79044 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P82930, Q9JIK9 | |
| MRPS34 | |
| Recombinant protein corresponding to Human MRPS34. Recombinant protein control fragment (Product #RP-98609). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction