missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS33 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93960-0.1ml
This item is not returnable.
View return policy
Description
MRPS33 Polyclonal antibody specifically detects MRPS33 in Human, Mouse samples. It is validated for Western Blot
Specifications
| MRPS33 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CGI-139, FLJ21123, mitochondrial ribosomal protein S33,28S ribosomal protein S33, mitochondrial, MRP-S33, PTD003, S33mt | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-106 of human MRPS33 (NP_057155.1). MSSLSEYAFRMSRLSARLFGEVTRPTNSKSMKVVKLFSELPLAKKKETYDWYPNHHTYAELMQTLRFLGLYRDEHQDFMDEQKRLKKLRGKEKPKKGEGKRAAKRK | |
| 0.1 mL | |
| Endocrinology | |
| 51650 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction