missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPS31 Recombinant Rabbit Monoclonal Antibody (3J3T5)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA555493
This item is not returnable.
View return policy
Description
Immunogen Sequence: NGFEELIQWT KEGKLWEFPI NNEAGFDDDG SEFHEHIFLE KHLESFPKQG PIRHFMELVT CGLSKNPYLS VKQKVEHIEW FRNYFNEKKD ILKESNIQFN.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. The 28S subunit of the mammalian mitoribosome may play a crucial and characteristic role in translation initiation. This gene encodes a 28S subunit protein that has also been associated with type 1 diabetes; however, its relationship to the etiology of this disease remains to be clarified. Pseudogenes corresponding to this gene have been found on chromosomes 3 and 13.
Specifications
| MRPS31 | |
| Recombinant Monoclonal | |
| 2 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| B0BN56, Q61733, Q92665 | |
| MRPS31 | |
| A synthetic peptide corresponding to a sequence within amino acids 296-395 of human MRPS31. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 3J3T5 | |
| Unconjugated | |
| MRPS31 | |
| 1500002D03Rik; 28S ribosomal protein S31, mitochondrial; AA960481; Imogen 38; Imogen38; Imogen44; Imogn38; islet mitochondrial antigen, 38 kD; mitochondrial ribosomal protein S31; Mitochondrial small ribosomal subunit protein mS31; Mrps31; MRP-S31; S31mt | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10240, 290850, 57312 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction