missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPS30 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576836
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Mitochondrial ribosomes consist of a large 39S subunit and a small 28S subunit, both of which are comprised of multiple mitochondrial ribosomal proteins (MRPs) that are encoded by nuclear genes and are essential for protein synthesis within mitochondria. MRP-S30 (mitochondrial ribosomal protein S30), also known as PDCD9 (programmed cell death protein 9), is a 439 amino acid protein that localizes to the mitochondrion, where it exists as a component of the 28S ribosomal subunit and works in conjunction with other MRPs to mediate protein synthesis. MRP-S30 is expressed in kidney, liver, heart and skeletal muscle. The gene encoding MRP-S30 maps to human chromosome 5, which contains 181 million base pairs and comprises nearly 6% of the human genome. Deletion of the p arm of chromosome 5 leads to Cri du chat syndrome, while deletion of the q arm or of chromosome 5 altogether is common in therapy-related acute myelogenous leukemias and myelodysplastic syndrome.
Specifications
| MRPS30 | |
| Polyclonal | |
| Unconjugated | |
| Mrps30 | |
| 2610020A16Rik; 28S ribosomal protein S30, mitochondrial; 39S ribosomal protein S30, mitochondrial; AA968347; BM-047; Mitochondrial large ribosomal subunit protein mL65; Mitochondrial large ribosomal subunit protein mS30; mitochondrial ribosomal protein S30; MRP S30; Mrps30; MRP-S30; PAP; PDCD9; programmed cell death 9; Programmed cell death protein 9; S30mt | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10884, 294767, 59054 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunocytochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9D0G0, Q9NP92 | |
| Mrps30 | |
| Recombinant full length Human MRPS30. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction