missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS27 Antibody (OTI1A2), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72784APCCY7
This item is not returnable.
View return policy
Description
MRPS27 Monoclonal specifically detects MRPS27 in Human samples. It is validated for Flow Cytometry.
Specifications
| MRPS27 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| MRPS27 | |
| Recombinant protein fragment corresponding to amino acids 151-414 of human MRPS27 (NP_055899) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| OTI1A2 | |
| Flow Cytometry | |
| FLJ21764, KIAA0264FLJ23348, mitochondrial, mitochondrial ribosomal protein S27, MRP-S27 | |
| Mouse | |
| Affinity Purified | |
| Stem Cell Markers | |
| 23107.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction