missing translation for 'onlineSavingsMsg'
Learn More

MRPS27 Antibody (1G3), Novus Biologicals™

Product Code. 18369868 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18369868 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18369868 Supplier Novus Biologicals Supplier No. H00023107M06

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MRPS27 Monoclonal antibody specifically detects MRPS27 in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPS27
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1G3
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH30521
Gene Alias FLJ21764, KIAA0264FLJ23348, mitochondrial, mitochondrial ribosomal protein S27, MRP-S27
Host Species Mouse
Immunogen MRPS27 (AAH30521, 51 a.a. ~ 168 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. SEEQSQNDEDNQGSEKLVEQLDIEETEQSKLPQYLERFKALHSKLQALGKIESEGLLSLTTQLVKEKLSTCEAEDIATYEQNLQQWHLDLVQLIQREQQQREQAKQEYQAQKAAKASA
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 23107
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.