missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MRPS26 Recombinant Rabbit Monoclonal Antibody (K01_1U07)

Product Code. 30827429
Change view
Click to view available options
Quantity:
50 μL
100 μL
Unit Size:
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30827429 50 μL 50µL
30826371 100 μL -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30827429 Supplier Invitrogen™ Supplier No. MA565052
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

MRPS26 Recombinant Monoclonal Antibody for ICC/IF, Western Blot

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein. This gene lies adjacent to and downstream of the gonadotropin-releasing hormone precursor gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPS26
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_1U07
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene MRPS26
Gene Accession No. Q9BYN8
Gene Alias 28S ribosomal protein S13, mitochondrial; 28S ribosomal protein S26, mitochondrial; 5'OT-EST protein; AI648866; C20orf193; dJ534B8.3; GI008; mitochondrial ribosomal protein S26; Mitochondrial small ribosomal subunit protein mS26; MRPS13; MRP-S13; Mrps26; MRP-S26; NY-BR-87; protein 5'OT-EST; RPMS13; rt26; S13mt; S26mt; serologically defined breast cancer antigen NY-BR-87
Gene Symbols MRPS26
Host Species Rabbit
Immunogen Recombinant protein of human MRPS26.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64949
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.