missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS26 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93351-0.02ml
This item is not returnable.
View return policy
Description
MRPS26 Polyclonal antibody specifically detects MRPS26 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| MRPS26 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:100-1:200, Immunohistochemistry-Paraffin | |
| C20orf193, dJ534B8.3, GI008,28S ribosomal protein S13, mitochondrial, mitochondrial ribosomal protein S26, MRP-S13MRPS13, MRP-S26chromosome 20 open reading frame 193, NY-BR-87, RPMS1328S ribosomal protein S26, mitochondrial, S13mt, S26mt, serologically defined breast cancer antigen NY-BR-87 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-205 of human MRPS26 (NP_110438.1). MLRALSRLGAGTPCRPRAPLVLPARGRKTRHDPLAKSKIERVNMPPAVDPAEFFVLMERYQHYRQTVRALRMEFVSEVQRKVHEARAGVLAERKALKDAAEHRELMAWNQAENRRLHELRIARLRQEEREQEQRQALEQARKAEEVQAWAQRKEREVLQLQEEVKNFITRENLEARVEAALDSRKNYNWAITREGLVVRPQRRDS | |
| 0.02 mL | |
| Endocrinology | |
| 64949 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction