missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS25 Antibody (3E6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00064432-M01
This item is not returnable.
View return policy
Description
MRPS25 Monoclonal antibody specifically detects MRPS25 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| MRPS25 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| FLJ00023, mitochondrial 28S ribosomal protein S25, mitochondrial ribosomal protein S25, MRP-S2528S ribosomal protein S25, mitochondrial, RPMS25DKFZp313H0817, S25mt | |
| MRPS25 (AAH03590, 1 a.a. ∽ 173 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MPMKGRFPIRRTLQYLSQGNVVFKDSVKVMTVNYNTHGELGEGARKFVFFNIPQIQYKNPWVQIMMFKNMTPSPFLRFYLDSGEQVLVDVETKSNKEIMEHIRKILGKNEETLREEEEEKKQLSHPANFGPRKYCLRECICEVEGQVPCPSLVPLPKEMRGKYKAALKADAQD | |
| 0.1 mg | |
| DNA replication Transcription Translation and Splicing | |
| 64432 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA | |
| 3E6 | |
| Western Blot 1:500, ELISA | |
| AAH03590 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction