missing translation for 'onlineSavingsMsg'
Learn More

MRPS25 Antibody (38E7), CoraFluor™ 1, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-42628CL1

Product Code. 30141725

  • £780.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MRPS25 Monoclonal antibody specifically detects MRPS25 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MRPS25
Monoclonal
CoraFluor 1
FLJ00023, mitochondrial 28S ribosomal protein S25, mitochondrial ribosomal protein S25, MRP-S2528S ribosomal protein S25, mitochondrial, RPMS25DKFZp313H0817, S25mt
Recombinant human MRPS25 (1-173aa) purified from E. coli
0.1 mL
DNA replication Transcription Translation and Splicing
64432
Store at 4°C in the dark. Do not freeze.
IgG2a κ
ELISA, Western Blot
38E7
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.