missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPS2 Monoclonal Antibody (OTI4D6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525318
This item is not returnable.
View return policy
Description
MRPS2 Monoclonal Antibody for Western Blot, IHC (P), Flow
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that belongs to the ribosomal protein S2 family. Alternatively spliced transcript variants have been observed for this gene.
Specifications
| MRPS2 | |
| Monoclonal | |
| 1.2 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9Y399 | |
| MRPS2 | |
| Full length human recombinant protein of MRPS2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI4D6 | |
| Unconjugated | |
| MRPS2 | |
| 1500019M10Rik; 28S ribosomal protein S2, mitochondrial; CGI-91; mitochondrial ribosomal protein S2; Mitochondrial small ribosomal subunit protein uS2m; MRPS2; MRP-S2; S2mt | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 51116 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction