missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPS18B Recombinant Rabbit Monoclonal Antibody (K01_2N37)
Description
MRPS18B Recombinant Monoclonal Antibody for ICC/IF, Western Blot
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that belongs to the ribosomal protein S18P family. The encoded protein is one of three that has significant sequence similarity to bacterial S18 proteins. The primary sequences of the three human mitochondrial S18 proteins are no more closely related to each other than they are to the prokaryotic S18 proteins. Pseudogenes corresponding to this gene are found on chromosomes 1q and 2q.
Specifications
Specifications
| Antigen | MRPS18B |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | K01_2N37 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 0.05% BSA, 150mM NaCl, 40% glycerol and 0.01% sodium azide; pH 7.4 |
| Gene | MRPS18B |
| Gene Accession No. | Q99N84, Q9Y676 |
| Gene Alias | 2400002C15Rik; 28S ribosomal protein S18-2, mitochondrial; 28S ribosomal protein S18b, mitochondrial; C6orf14; HSPC183; HumanS18a; mitochondrial ribosomal protein S18-2; mitochondrial ribosomal protein S18B; Mitochondrial small ribosomal subunit protein bS18b; Mitochondrial small ribosomal subunit protein mS40; MRP-S18-2; MRPS18-2; MRPS18B; MRP-S18-b; mrps18-b; PTD017; S18amt; S18mt-b |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?