missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MRPS18B Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA588338

Product Code. 16556611

  • £449.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MAASVLNTVL RRLPMLSLFR GSHRVQVPLQ TLCTKAPSEE DSLSSVPISP YKDEPWKYLE SEEYQERYGS RPVWADYRRN HKGGVPPQRT RKTCIRRNKV VGNPCPICRD HKLHVDFRNV KLLEQFVCAH TGIIFYAPYT GVCVKQHKRL TQAIQKARDH GLLIYHIPQV EPRDLDFSTS HGAVSATPPA PTLVSGDPWY PWYNWKQPPE RELSRLRRLY QGHLQEESGP PPESMPKMPP RTPAEASSTG QTGPQSAL; Positive Samples: U-87MG, A-431, Mouse kidney, Mouse brain, Rat heart, Rat kidney; Cellular Location: Mitochondrion.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that belongs to the ribosomal protein S18P family. The encoded protein is one of three that has significant sequence similarity to bacterial S18 proteins. The primary sequences of the three human mitochondrial S18 proteins are no more closely related to each other than they are to the prokaryotic S18 proteins. Pseudogenes corresponding to this gene are found on chromosomes 1q and 2q.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MRPS18B
Polyclonal
Unconjugated
MRPS18B
2400002C15Rik; 28S ribosomal protein S18-2, mitochondrial; 28S ribosomal protein S18b, mitochondrial; C6orf14; HSPC183; HumanS18a; mitochondrial ribosomal protein S18-2; mitochondrial ribosomal protein S18B; Mitochondrial small ribosomal subunit protein bS18b; Mitochondrial small ribosomal subunit protein mS40; MRP-S18-2; MRPS18-2; MRPS18B; MRP-S18-b; mrps18-b; PTD017; S18amt; S18mt-b
Rabbit
Affinity Chromatography
RUO
28973, 294230, 66973
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
4.53 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Q99N84, Q9Y676
MRPS18B
Recombinant fusion protein containing a sequence corresponding to amino acids 1-258 of human MRPS18B (NP_0547651).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.