missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS18B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-48958
This item is not returnable.
View return policy
Description
MRPS18B Polyclonal antibody specifically detects MRPS18B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| MRPS18B | |
| Polyclonal | |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| 28S ribosomal protein S18-2, mitochondrial, C6orf1428S ribosomal protein S18b, mitochondrial, HSPC183, HumanS18a, mitochondrial ribosomal protein S18-2, mitochondrial ribosomal protein S18B, MRP-S18-2, MRPS18-2DKFZp564H0223, Mrps18-b, MRP-S18-b, PTD017, S18amt, S18mt-b | |
| This antibody was developed against a recombinant protein corresponding to amino acids: ASVLNTVLRRLPMLSLFRGSHRVQVPLQTLCTKAPSEEDSLSSVPISPYKDEPWKYLESEEYQERYGSRPVWADYRRNHKGGVPPQRTRK | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 28973 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction