missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS18B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35212-100ul
This item is not returnable.
View return policy
Description
MRPS18B Polyclonal antibody specifically detects MRPS18B in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| MRPS18B | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| 28S ribosomal protein S18-2, mitochondrial, C6orf1428S ribosomal protein S18b, mitochondrial, HSPC183, HumanS18a, mitochondrial ribosomal protein S18-2, mitochondrial ribosomal protein S18B, MRP-S18-2, MRPS18-2DKFZp564H0223, Mrps18-b, MRP-S18-b, PTD017, S18amt, S18mt-b | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-258 of human MRPS18B (NP_054765.1).,, Sequence:, MAASVLNTVLRRLPMLSLFRGSHRVQVPLQTLCTKAPSEEDSLSSVPISPYKDEPWKYLESEEYQERYGSRPVWADYRRNHKGGVPPQRTRKTCIRRNKVVGNPCPICRDHKLHVDFRNVKLLEQFVCAHTGIIFYAPYTGVCVKQHKRLTQAIQKARDHGLLIYHIPQVEPRDLDFSTSHGAVSATPPAPTLVSGDPWYPWYNWKQPPERELSRLRRLYQGHLQEESGPPPESMPKMPPRTPAEASSTGQTGPQSAL | |
| 100 μL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 28973 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction