missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MRPS18A Polyclonal antibody specifically detects MRPS18A in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | MRPS18A |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 532 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | 28S ribosomal protein S18a, mitochondrial, FLJ10548, mitochondrial, mitochondrial ribosomal protein S18-3, mitochondrial ribosomal protein S18A, MRP-S18-3, MRPS18-3HumanS18b, Mrps18a, MRP-S18-a, S18bmt, S18mt-a |
| Host Species | Rabbit |
| Immunogen | mitochondrial ribosomal protein S18A. (Uniprot# Q9NVS2) |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?