missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS17 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18881-1-AP-20UL
This item is not returnable.
View return policy
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that belongs to the ribosomal protein S17P family. The encoded protein is moderately conserved between human mitochondrial and prokaryotic ribosomal proteins. Pseudogenes corresponding to this gene are found on chromosomes 1p, 3p, 6q, 14p, 18q, and Xq.Specifications
| MRPS17 | |
| Polyclonal | |
| Unconjugated | |
| MRPS17 | |
| HSPC011, MRP S17, MRPS17, RPMS17, S17mt | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 288621, 51373, 66258 | |
| -20°C | |
| Liquid |
| Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9CQE3, Q9Y2R5 | |
| Mrps17 | |
| MRPS17 Fusion Protein Ag13414 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction