Learn More
Invitrogen™ MRPS16 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597277
Description
Immunogen sequence: MVHLTTLLCK AYRGGHLTIR LALGGCTNRP FYRIVAAHNK CPRDGRFVEQ LGSYDPLPNS HGEKLVALNL DRIRHWIGCG AHLSKPMEKL LGLAGFFPLH PMMITNAERL RRKRAREVLL ASQKTDAEAT DTEATET; Positive Samples: HeLa, Jurkat, SW620, HT-1080, MCF7, Mouse heart, Mouse kidney, Mouse liver, Rat brain; Cellular Location: Mitochondrion.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that belongs to the ribosomal protein S16P family. The encoded protein is one of the most highly conserved ribosomal proteins between mammalian and yeast mitochondria. Three pseudogenes (located at 8q21. 3, 20q13. 32, 22q12-q13. 1) for this gene have been described.
Specifications
| MRPS16 | |
| Polyclonal | |
| Unconjugated | |
| Mrps16 | |
| 1500011E11Rik; 28S ribosomal protein S16, mitochondrial; CGI-132; COXPD2; mitochondrial ribosomal protein S16; Mitochondrial small ribosomal subunit protein bS16m; Mprs16; Mrps16; MRP-S16; Rpms16; S16mt | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 51021, 66242, 688912 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.80 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9CPX7, Q9Y3D3 | |
| Mrps16 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.