missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MRPS16 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15729931 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15729931 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15729931 Supplier Invitrogen™ Supplier No. PA562131

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: ALNLDRIRHW IGCGAHLSKP MEKLLGLAGF FPLHPMMITN AERLRRKRAR EVLLASQKTD AEATDT Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 88%.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that belongs to the ribosomal protein S16P family. The encoded protein is one of the most highly conserved ribosomal proteins between mammalian and yeast mitochondria. Three pseudogenes (located at 8q21. 3, 20q13. 32, 22q12-q13. 1) for this gene have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPS16
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Mrps16
Gene Accession No. Q9Y3D3
Gene Alias 1500011E11Rik; 28S ribosomal protein S16, mitochondrial; CGI-132; COXPD2; mitochondrial ribosomal protein S16; Mitochondrial small ribosomal subunit protein bS16m; Mprs16; Mrps16; MRP-S16; Rpms16; S16mt
Gene Symbols Mrps16
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MRPS16. Recombinant protein control fragment (Product #RP-100646).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51021
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.