missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MRPS15 Monoclonal antibody specifically detects MRPS15 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | MRPS15 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI6D2 |
| Conjugate | Alexa Fluor 350 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | FLJ11564,28S ribosomal protein S15, mitochondrial, mitochondrial ribosomal protein S15, MPR-S15, MRP-S15, RPMS15, S15mt |
| Host Species | Mouse |
| Immunogen | Full length human recombinant protein of human MRPS15(NP_112570) produced in E.coli. |
| Purification Method | >95% |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?