missing translation for 'onlineSavingsMsg'
Learn More

MRPS15 Antibody (OTI5F5) [Biotin], Novus Biologicals™

Product Code. 30688809 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30688809 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30688809 Supplier Novus Biologicals Supplier No. NBP344911B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MRPS15 Monoclonal antibody specifically detects MRPS15 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPS15
Applications Western Blot
Classification Monoclonal
Clone OTI5F5
Conjugate Biotin
Formulation PBS
Gene Alias FLJ11564,28S ribosomal protein S15, mitochondrial, mitochondrial ribosomal protein S15, MPR-S15, MRP-S15, RPMS15, S15mt
Host Species Mouse
Immunogen Full length human recombinant protein of human MRPS15(NP_112570) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64960
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.