Learn More
Invitrogen™ MRPS14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120370
Description
Positive test controls include: BxPC-3, U-251MG, Mouse heart, Mouse kidney, Mouse liver. The target is usually found in the following locations: Mitochondrion. Immunogen sequence: MAAFMLGSLL RTFKQMVPSS ASGQVRSHYV DWRMWRDVKR RKMAYEYADE RLRINSLRKN TILPKILQDV ADEEIAALPR DSCPVRIRNR CVMTSRPRGV KRRWRLSRIV FRHLADHGQL SGIQRATW.
Specifications
| MRPS14 | |
| Polyclonal | |
| Unconjugated | |
| MRPS14 | |
| 1810032L21Rik; 28S ribosomal protein S14, mitochondrial; AL022847; DJ262D12.2; HSMRPS14; mitochondrial 28S ribosomal protein S14; mitochondrial ribosomal protein S14; Mitochondrial small ribosomal subunit protein uS14m; MRPS14; MRP-S14; Rpms14; S14mt | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 63931, 64659 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.1 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O60783, Q9CR88 | |
| MRPS14 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-128 of human MRPS14 (NP_071383.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.