missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS14 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93952-0.1ml
This item is not returnable.
View return policy
Description
MRPS14 Polyclonal antibody specifically detects MRPS14 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| MRPS14 | |
| Polyclonal | |
| Western Blot 1:200-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| DJ262D12.2, HSMRPS14, mitochondrial 28S ribosomal protein S14, mitochondrial ribosomal protein S14,28S ribosomal protein S14, mitochondrial, MRP-S14, S14mt | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-128 of human MRPS14 (NP_071383.1). MAAFMLGSLLRTFKQMVPSSASGQVRSHYVDWRMWRDVKRRKMAYEYADERLRINSLRKNTILPKILQDVADEEIAALPRDSCPVRIRNRCVMTSRPRGVKRRWRLSRIVFRHLADHGQLSGIQRATW | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 63931 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction