missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS12 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA538947
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that belongs to the ribosomal protein S12P family. The encoded protein is a key component of the ribosomal small subunit and controls the decoding fidelity and susceptibility to aminoglycoside antibiotics. The gene for mitochondrial seryl-tRNA synthetase is located upstream and adjacent to this gene, and both genes are possible candidates for the autosomal domit deafness gene (DFNA4). Splice variants that differ in the 5′ UTR have been found for this gene; all three variants encode the same protein. [provided by RefSeq, Jul 2008].
Especificaciones
| MRPS12 | |
| Polyclonal | |
| Unconjugated | |
| MRPS12 | |
| RPMS12, RPSM12 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 24030, 6183 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O15235, O35680 | |
| MRPS12 | |
| A synthetic peptide derived from the internal region of human MRPS12 | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido